WHA67033650
Whatman® Polycap HD heavy duty encapsulated filter
Polycap HD 36, pore size 5 μm
Synonym(s):
Whatman filter, Whatman filters, Z743674
About This Item
Recommended Products
material
polypropylene housing
polypropylene membrane
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Whatman 6703-3650
Whatman Article No. 28420120 (US reference)
diam.
36 mm
pore size
5 μm
fitting
inlet Stepped barb for 6 to 10 mm (1/4 to 3/8 in.) tubing
outlet Stepped barb for 6 to 10 mm (1/4 to 3/8 in.) tubing
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Heavy duty polypropylene filter capsule for chemical processing
Whatman Polycap HD capsule filters are used in processing applications to filter buffers, solvents, reagents and emulsions across a wide range of industries including clean air and gas equipment, cosmetics and personal care, food and beverage, inks and pigments, pharmaceuticals, photography, semiconductors and magnetic media.
Chemical processing applications often require a capsule filter to ensure a finishing step that yields a high purity final product. Different solution temperatures and pH values can complicate filtration. Whatman Polycap HD capsule filters are manufactured with polypropylene filter media, support system and housing allow use with a broad range of solutions, pH, and temperatures. Sturdy construction enables high flow rate and high retention capacity for excellent filtrate purity.
Features and Benefits
- Can be autoclaved with steam at 121°C for 20 min
- Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
- Available in 0.45, 1.0, 5.0, 10, or 20 µm pore sizes, as well as a variety of end-fitting configurations
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service