WHA10414112
Whatman® polyamide NL17 filter discs 0.45 μm pore size
white, hydrophilic, 100 ea, 47 mm diam
Synonym(s):
Z134341, membrane filters, nylon membranes
About This Item
Recommended Products
Product Name
Whatman® polyamide membrane filters, polyamide membrane circles, (NL17), 0.45 μm, 47 mm, 100/pk
material
Nylon membrane (polyamide membrane)
sterility
non-sterile
packaging
pack of 100
manufacturer/tradename
Whatman 10414112
Whatman Article No. 28415338 (US reference)
diam.
47.0 mm
pore size
0.45 μm pore size
suitability
suitable for (clarification and sterile filtration.)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
NL 16
- Pore size (μm): 0.2 (NL 16)
- Thickness (μm): 110
- Nominal pore size (μm): 0.2
- Water flow rate Δp = 0.9 bar (mL/min/cm2): 10
- Bubble point (bar): 4.2
- Maximum temperature: 135°C
NL 17
- Pore size (μm): 0.45 (NL 17)
- Thickness (μm): 110
- Nominal pore size (μm): 0.45
- Water flow rate Δp = 0.9 bar (mL/min/cm2): 20
- Bubble point (bar): 2.8
- Maximum temperature: 135°
Features and Benefits
- They are hydrophilic making them suitable for aqueous and organic solutions.
- The membrane filters can be used up to 135°C.
- Applications include: filtration of aqueous and organic mobile phases, vacuum degassing, filtration of tissue culture media, microbiological media, buffers, and solutions.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service