WHA10347670
Whatman® weighing boats and paper
B-2 Sheets, dish L × W 12 mm × 304 mm
Synonym(s):
Z696323, weighing boats, weighing paper
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.71
Recommended Products
material
paper
feature
grade B-2
packaging
pack of 500 ea
manufacturer/tradename
Whatman 10347670
Whatman Article No. 28414661 (US reference)
dish L × W
12 mm × 304 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Grade B-2 Whatman weighing paper from Cytiva dissolves residue-free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
Packs of 500 Grade B-2 Whatman weighing paper are available in four sheet sizes:
Packs of 500 Grade B-2 Whatman weighing paper are available in four sheet sizes:
- 76.2 × 76.2 mm (3 × 3 inch) sheets
- 101.6 × 101.6 mm (4 × 4 inch) sheets
- 152.4 × 152.4 mm (6 × 6 inch) sheets
- 304.8 × 304.8 mm (12 × 12 inch) sheets
Features and Benefits
- Thin (0.04 mm) and lightweight (43 g/m²)
- Transparent and smooth parchment surface
- Simple weighing out and sample transfer
- Reliable for quantitative transfer
- Range of sheet sizes for ease-of-use
- Low nitrogen paper that is residue-free for accurate chemical analysis
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service