WHA10312544
Whatman® qualitative filter paper, Grade 602eh 1/2
circles, diam. 125 mm, pack of 100
Synonym(s):
Z749257
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Recommended Products
material
cellulose filters
circles
packaging
pack of 100
manufacturer/tradename
Whatman 10312544
Whatman Article No. 28414276 (US reference)
parameter
3000 sec/100 mL speed (Herzberg)
diam.
125 mm
pore size
2 μm
basis weight
85 g/m2
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Grade 602EH½ Qualitative Filter Papers for very fine precipitates.
Cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters give improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.
Cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters give improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.
Features and Benefits
- Maintains flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
- Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
- Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
- Increased total loading capacity as more filter area is available.
- The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service