Skip to Content
Merck
All Photos(1)

Key Documents

W511609

Sigma-Aldrich

2-Methylquinoxaline

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
Beilstein:
113307
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.139
NACRES:
NA.21
Organoleptic:
coffee; nutty; roasted
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

Assay

≥97%

refractive index

n20/D 1.613 (lit.)

bp

245-247 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; nutty; roasted

SMILES string

Cc1cnc2ccccc2n1

InChI

1S/C9H8N2/c1-7-6-10-8-4-2-3-5-9(8)11-7/h2-6H,1H3

InChI key

ALHUXMDEZNLFTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Methylquinoxaline is a heterocyclic compound used in roasted and meat flavoring. It is one of the volatile compounds formed during the thermal reaction between L-cysteine and dihydroxyacetone in glycerine.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Rowe DJ
Chemistry and Technology of Flavors and Fragrances., 110-110 (2004)
F W Chaplen et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(10), 5533-5538 (1998-05-20)
Methylglyoxal is an alpha-ketoaldehyde and dicarbonyl formed in cells as a side product of normal metabolism. Endogenously produced dicarbonyls, such as methylglyoxal, are involved in numerous pathogenic processes in vivo, including carcinogenesis and advanced glycation end-product formation; advanced glycation end-products
Volatile Products Formed from L-Cysteine and Dihydroxyacetone Thermally Treated in Different Solvents.
Okumura J, et al.
Agricultural and Biological Chemistry, 54(7), 1631-1638 (1990)
Determination of methylglyoxal as 2-methylquinoxaline by high-performance liquid chromatography and its application to biological samples.
S Ohmori et al.
Journal of chromatography, 414(1), 149-155 (1987-02-20)
Methylglyoxal assay in cells as 2-methylquinoxaline using 1,2-diaminobenzene as derivatizing reagent.
C Cordeiro et al.
Analytical biochemistry, 234(2), 221-224 (1996-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service