Skip to Content
Merck
All Photos(2)

Key Documents

W409901

Sigma-Aldrich

2,4-Dimethyl-1,3-dioxolane

≥98%, FG

Synonym(s):

Propylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
FEMA Number:
4099
MDL number:
UNSPSC Code:
12164502
Flavis number:
06.077
NACRES:
NA.21

grade

FG
Fragrance grade

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

grassy; green; fruity

InChI

1S/C5H10O2/c1-4-3-6-5(2)7-4/h4-5H,3H2,1-2H3

InChI key

ROSFUFIOLRQOON-UHFFFAOYSA-N

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service