W397025
2,5-Dimethyl-3-oxo-(2H)-fur-4-yl butyrate
natural, ≥93%
Synonym(s):
Furanone butyrate, Strawberry furanone butyrate
About This Item
Recommended Products
grade
Halal
Kosher
natural
Quality Level
Agency
meets purity specifications of JECFA
reg. compliance
FDA 21 CFR 117
Assay
≥93%
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
greener alternative category
Organoleptic
caramel; brown; sweet
storage temp.
2-8°C
InChI
1S/C10H14O4/c1-4-5-8(11)14-10-7(3)13-6(2)9(10)12/h6H,4-5H2,1-3H3
InChI key
JOLGAERKCZYHLF-UHFFFAOYSA-N
Related Categories
General description
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service