Skip to Content
Merck
All Photos(3)

Key Documents

W379601

Sigma-Aldrich

Vanillyl butyl ether

greener alternative

natural (US), 97%, FG

Synonym(s):

4-(butoxymethyl)-2-methoxy-Phenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
210.27
FEMA Number:
3796
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.093
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

97%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.516 (lit.)

bp

241 °C (lit.)

solubility

soluble (Insoluble in water. Soluble in organic solvents, oils.)

density

1.057 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

creamy; sweet; vanilla

SMILES string

CCCCOCC1=CC(OC)=C(O)C=C1

InChI

1S/C12H18O3/c1-3-4-7-15-9-10-5-6-11(13)12(8-10)14-2/h5-6,8,13H,3-4,7,9H2,1-2H3

InChI key

VLDFMKOUUQYFGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vanillyl butyl ether is an alkoxy-substituted benzyl derivative mainly used as a flavoring agent.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product is a Biobased product aligning with the principles of "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks" in Green Chemistry.

Application

This compound has a characteristic trigeminal, burning, hot pepper nature and can be used in spice flavors like pepper, cinnamon and ginger, as well as blends for some baked applications, including cookies. Other potential applications include flavoring for alcoholic and non-alcoholic beverages with herbal, berry, vanilla, cola, ginger ale, root beer, hot chocolate, chai, ice tea and mints.

Biochem/physiol Actions

Taste at 10-15%

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam TB, et al.
Adam TB, et al.
Food And Chemical Toxicology, 43(8), 1241-1271 (2005)
Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(4), 50-50 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service