Skip to Content
Merck
All Photos(1)

Key Documents

W360600

Sigma-Aldrich

D-(+)-Xylose

≥99%

Synonym(s):

(2S,3R,4S,5R)-oxane-2,3,4,5-tetrol, Dextro-xylose, Wood sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
FEMA Number:
3606
Beilstein:
1562108
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥99%

optical activity

[α]20/D +18.8°, c = 4 in H2O

mp

154-158 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

smoky

SMILES string

O[C@@H]1COC(O)[C@H](O)[C@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4-,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-IOVATXLUSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Smoky

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shmuel Gleizer et al.
Cell, 179(6), 1255-1263 (2019-11-30)
The living world is largely divided into autotrophs that convert CO2 into biomass and heterotrophs that consume organic compounds. In spite of widespread interest in renewable energy storage and more sustainable food production, the engineering of industrially relevant heterotrophic model
Nunzia D'Urzo et al.
Microbial cell factories, 12, 12-12 (2013-02-05)
In past years research has focused on the development of alternative Gram positive bacterial expression systems to produce industrially relevant proteins. Brevibacillus choshinensis is an easy to handle non-sporulating bacterium, lacking extracellular proteases, that has been already shown to provide
Thu V Vuong et al.
PloS one, 9(4), e95170-e95170 (2014-04-17)
The xylan-binding module Clostridium thermocellum CBM22A was successfully fused to a gluco-oligosaccharide oxidase, GOOX-VN, from Sarocladium strictum via a short TP linker, allowing the fused protein to effectively bind different xylans. The presence of the CtCBM22A at the N-terminal of
Heather A Feaga et al.
mBio, 5(6), e01916-e01916 (2014-11-13)
Bacterial ribosomes frequently translate to the 3' end of an mRNA without terminating at a stop codon. Almost all bacteria use the transfer-messenger RNA (tmRNA)-based trans-translation pathway to release these "nonstop" ribosomes and maintain protein synthesis capacity. trans-translation is essential
Jose Serate et al.
Biotechnology for biofuels, 8, 180-180 (2015-11-20)
Microbial conversion of lignocellulosic feedstocks into biofuels remains an attractive means to produce sustainable energy. It is essential to produce lignocellulosic hydrolysates in a consistent manner in order to study microbial performance in different feedstock hydrolysates. Because of the potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service