Skip to Content
Merck
All Photos(1)

Key Documents

W339008

Sigma-Aldrich

Fenchyl acetate, mixture of α- and β-

≥96%, FG

Synonym(s):

1,3,3-Trimethyl-2-norbornanyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
3390
EC Number:
Council of Europe no.:
11769
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.269
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥96%

optical activity

[α]22/D +54°, neat

refractive index

n20/D 1.456 (lit.)

bp

220 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsamic; pine; sweet

SMILES string

CC(=O)OC1C(C)(C)[C@@H]2CC[C@@]1(C)C2

InChI

1S/C12H20O2/c1-8(13)14-10-11(2,3)9-5-6-12(10,4)7-9/h9-10H,5-7H2,1-4H3/t9-,10?,12+/m1/s1

InChI key

JUWUWIGZUVEFQB-MAZPRZIYSA-N

Related Categories

General description

Fenchyl acetate is a fragrance ingredient found in many fragrance compounds including fine fragrances, decorative cosmetics, shampoos, toilet soaps, and other toiletries .

Application

Fenchyl acetate, a mixture of α- and β- contributes to the overall aroma and flavor profile of Alpinia calcarata .

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on 1, 3, 3-trimethyl-2-norbornanyl acetate
Bhatia SP, et al.
Food And Chemical Toxicology, 46(12), S106-S108 (2008)
Chemical diversity in the genus Alpinia (Zingiberaceae): comparative composition of four Alpinia species grown in Northern India
Padalia RC, et al.
Chemistry and Biodiversity, 7(8), 2076-2087 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service