Skip to Content
Merck
All Photos(1)

Key Documents

W331715

Sigma-Aldrich

2-Pentylfuran

natural (US), ≥97%, FG

Synonym(s):

2-Amylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21
FEMA Number:
3317
EC Number:
Council of Europe no.:
10966
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.059
NACRES:
NA.21

grade

FG
Fragrance grade
Kosher
natural (US)

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.448 (lit.)

bp

64-66 °C/23 mmHg (lit.)

density

0.883 g/mL at 20 °C (lit.)
0.886 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; earthy; vegetable; fruity

SMILES string

CCCCCc1ccco1

InChI

1S/C9H14O/c1-2-3-4-6-9-7-5-8-10-9/h5,7-8H,2-4,6H2,1H3

InChI key

YVBAUDVGOFCUSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Pentylfuran is a volatile heterocyclic compound reported to be the responsible for the undesirable reversion flavor of soybean oil. It may be formed by the autoxidation of linoleic acid.
Color: may darken over time

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Analysis of autoxidized fats by gas chromatography-mass spectrometry: VII. Volatile thermal decomposition products of pure hydroperoxides from autoxidized and photosensitized oxidized methyl oleate, linoleate and linolenate.
Frankel EN, et al.
Lipids, 16(5), 279-285 (1981)
Isolation and identification of 2-pentyl-furan as contributing to the reversion flavour of soyabean oil
Chang SS, et al.
Chemistry & Industry, 46, 1926-1926 (1966)
Singlet Oxygen Oxidation for 2?Pentylfuran and 2?Pentenyfuran Formation in Soybean Oil.
Min DB, et al.
Journal of Food Science, 68(4), 1175-1178 (2003)
Identification of 2?Pentyl Furan in Fats and Oils and Its Relationship to the Reversion Flavor of Soybean Oil.
Krishnamurthy RG, et al.
Journal of Food Science, 32(4), 372-374 (1967)
A systematic characterization of the reversion flavor of soybean oil.
Smouse TH & Chang SS
Journal of the American Oil Chemists' Society, 44(8), 509-514 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service