W331007
Methyl thiobutyrate
≥97%, FG
Synonym(s):
S-Methyl thiobutanoate, S-Methyl thiobutyrate
About This Item
Fragrance grade
Halal
Kosher
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
Assay
≥97%
refractive index
n20/D 1.461 (lit.)
bp
142-143 °C/757 mmHg (lit.)
density
0.966 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
cheese; musty; sulfurous
SMILES string
CCCC(=O)SC
InChI
1S/C5H10OS/c1-3-4-5(6)7-2/h3-4H2,1-2H3
InChI key
GRLJIIJNZJVMGP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- RIFM fragrance ingredient safety assessment, methyl thiobutyrate, CAS Registry Number 2432-51-1.: This study reviews the safety of methyl thiobutyrate as a fragrance ingredient, evaluating its toxicity and potential effects in various applications (Api et al., 2020).
- Carboxylesterase activity, cDNA sequence, and gene expression in malathion susceptible and resistant strains of the cotton aphid, Aphis gossypii.: The study examines the role of carboxylesterases in pesticide resistance, with methyl thiobutyrate used to characterize enzyme activity (Pan et al., 2009).
- Odorant and pheromone binding by aphrodisin, a hamster aphrodisiac protein.: Research on aphrodisin′s binding properties includes the evaluation of methyl thiobutyrate as a potential ligand, contributing to the understanding of pheromone signaling mechanisms (Briand et al., 2000).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
93.2 °F - closed cup
Flash Point(C)
34 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service