Skip to Content
Merck
All Photos(1)

Key Documents

W329800

Sigma-Aldrich

3-Mercapto-2-butanone

≥95%, FG

Synonym(s):

3-sulfanylbutan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(SH)COCH3
CAS Number:
Molecular Weight:
104.17
FEMA Number:
3298
EC Number:
Council of Europe no.:
11497
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.047
NACRES:
NA.21
Organoleptic:
meaty; onion; roasted; sulfurous
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥95%

bp

48-49 °C/15 mmHg (lit.)

density

1.035 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; onion; roasted; sulfurous

storage temp.

2-8°C

SMILES string

CC(S)C(C)=O

InChI

1S/C4H8OS/c1-3(5)4(2)6/h4,6H,1-2H3

InChI key

XLMPYCGSRHSSSX-UHFFFAOYSA-N

Related Categories

General description

3-Mercapto-2-butanone is a sulfur-containing flavor compound mainly found in meat and meat products. It is formed by the Maillard reaction between sugars and cysteine or thiamin, which are known precursors for meat-like flavorings.

Biochem/physiol Actions

Taste at 0.3-1.0 ppm

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

117.0 °F - closed cup

Flash Point(C)

47.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aroma volatiles from meatlike Maillard systems.
Mottram DS & Whitfield FB.
ACS Symp. Ser., 543, 180?191-180?191 (1994)
Origin of carbons in sulfur-containing aroma compounds from the Maillard reaction of xylose, cysteine and thiamine
Cerny C.
LWT--Food Science and Technology, 40(8), 1309-1315 (2007)
Important sulfur-containing aroma volatiles in meat.
Mottram DS & Madruga MS
ACS Symp. Ser., 564, 180?187-180?187 (1994)
Flavour formation in meat and meat products: a review.
Mottram DS
Food Chemistry, 62(4), 415-424 (1998)
Mosciano, G.
Perfumer & Flavorist, 26, 70-70 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service