Skip to Content
Merck
All Photos(1)

Key Documents

W315605

Sigma-Aldrich

4-Ethylphenol

≥98%, FG

Synonym(s):

1-ethyl-4-hydroxybenzene, 4-hydroxyphenyl ethane, p-ethylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4OH
CAS Number:
Molecular Weight:
122.16
FEMA Number:
3156
Beilstein:
1363317
EC Number:
Council of Europe no.:
550
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39023356
PubChem Substance ID:
Flavis number:
4.022
NACRES:
NA.21
Organoleptic:
smoky; phenolic
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

vapor density

4.2 (vs air)

vapor pressure

0.13 mmHg ( 20 °C)

Assay

≥98%

bp

218-219 °C (lit.)

mp

40-42 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

smoky; phenolic

SMILES string

CCc1ccc(O)cc1

InChI

1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3

InChI key

HXDOZKJGKXYMEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Ethylphenol (4-EP) is a para-substituted phenolic compound. It is one of the important aroma compounds in red wine.[1] The aroma associated with 4-EP in red wine has been described as ′′horsy′′, ′′leather′′, ′′medicinal′′, ′′smoky′′, ′′barnyard′′, ′′animal′′ and ′′sweaty saddle′′-like.[1] Production of 4-EP from p-coumaric acid in synthetic media utilizing several yeast species associated with wine production has been reported.[2]

Application


  • Sorption Behavior of Volatile Phenols at the Oak Wood/Wine Interface in a Model System.: This research explores the interaction between 4-ethylphenol and oak wood in wine aging, contributing to the understanding of volatile phenol adsorption and its impact on wine flavor (Barrera-García et al., 2006).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of 4-Ethylphenol and 4-Ethylguaiacol with Polyaniline-Based Compounds in Wine-Like Model Solutions and Red Wine.
Carrasco-Sanchez V, et al.
Molecules (Basel), 20(8), 14312-14325 (2015)
Phenol and para-substituted phenols electrochemical oxidation pathways.
Enache TA and Oliveira-Brett AM.
Journal of Electroanalytical Chemistry, 655(1), 9-16 (2011)
W McIntire et al.
The Biochemical journal, 224(2), 617-621 (1984-12-01)
Enzymic hydroxylation of 4-ethylphenol by (a) Pseudomonas putida and (b) highly purified p-cresol methylhydroxylase gave optically active 1-(4'-hydroxyphenyl)-ethanol. The products were transformed into the phenolic methyl ethers and shown to contain 69.5% and 65.6%, respectively, of the (S)-(-)-isomer. The stereochemistry
Identification of yeasts isolated from wine-related environments and capable of producing 4-ethylphenol.
Dias L, et al.
Food Microbiology, 20(5), 567-574 (2003)
Factors affecting the production of 4-ethylphenol by the yeast Dekkera bruxellensis in enological conditions.
Dias L, et al.
Food Microbiology, 20(4), 377-384 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service