W279617
γ-Octalactone
natural, ≥97%, FG
Synonym(s):
γ-Octanoic lactone, 4-Hydroxy octanoic acid, Octano-1,4-lactone
About This Item
Fragrance grade
Halal
Kosher
natural
meets purity specifications of JECFA
Recommended Products
grade
FG
Fragrance grade
Halal
Kosher
natural
Quality Level
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515
Assay
≥97%
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
refractive index
n20/D 1.444 (lit.)
bp
234 °C (lit.)
density
0.981 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
greener alternative category
Organoleptic
coconut; creamy; peach; sweet
SMILES string
CCCCC1CCC(=O)O1
InChI
1S/C8H14O2/c1-2-3-4-7-5-6-8(9)10-7/h7H,2-6H2,1H3
InChI key
IPBFYZQJXZJBFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Effect of osmotic dehydration combined with vacuum freeze-drying treatment on characteristic aroma components of peach slices.: This study explores the impact of combined osmotic dehydration and vacuum freeze-drying on the aroma components of peach slices. γ-Octalactone was identified as a key compound contributing to the enhanced aroma profile, providing insights into improving the sensory qualities of dehydrated fruits (Wang et al., 2024).
- The Application of Synthetic Flavors in Zebrafish (Danio rerio) Rearing with Emphasis on Attractive Ones: Effects on Fish Development, Welfare, and Appetite.: This research investigates the use of synthetic flavors, including γ-Octalactone, in zebrafish rearing. The study found that γ-Octalactone significantly improved fish development, welfare, and appetite, highlighting its potential in aquaculture applications (Conti et al., 2023).
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service