Skip to Content
Merck
All Photos(1)

Key Documents

W261602

Sigma-Aldrich

Lauryl acetate

≥98%, FG

Synonym(s):

Dodecyl acetate, Lauryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2(CH2)11CH3
CAS Number:
Molecular Weight:
228.37
FEMA Number:
2616
EC Number:
Council of Europe no.:
200
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.010
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.432 (lit.)

bp

150 °C/15 mmHg (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; waxy; sweet

SMILES string

CCCCCCCCCCCCOC(C)=O

InChI

1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-16-14(2)15/h3-13H2,1-2H3

InChI key

VZWGRQBCURJOMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth M MacDonald et al.
Journal of chemical ecology, 29(10), 2385-2389 (2003-12-20)
Larvae of the western flower thrips Frankliniella occidentalis are known to produce an anal droplet containing decyl acetate (10Ac) and dodecyl acetate (12Ac), which act as an alarm pheromone. Analysis by GC showed that the combined mass of 10Ac and
Ido Yosha et al.
Journal of agricultural and food chemistry, 56(17), 8045-8049 (2008-08-12)
Alginate-gelatin beads with dispersed droplets of a model pheromone, dodecyl acetate, were prepared as a vehicle for slow release of pheromones into the atmosphere over a prolonged period of time. The beads are prepared in two steps, the first being
K D Philipson et al.
The American journal of physiology, 248(1 Pt 2), H147-H150 (1985-01-01)
We have used charged amphiphiles as phospholipid analogues to modulate the interaction of Ca2+ with myocardial sarcolemma. The amphiphiles were dodecyl sulfate, dodecyltrimethylamine, and lauryl acetate; these are anionic, cationic, and neutral molecules, respectively. The hydrophobic alkyl chain is identical
Developing new anti-cancer drugs.
Ricardo de Souza Pereira
Recent patents on anti-cancer drug discovery, 6(1), 1-5 (2010-11-30)
Brent L Waguespack et al.
Journal of chromatography. A, 1078(1-2), 171-180 (2005-07-13)
A variety of porous polymer monoliths (PPMs) have been synthesized using the 'conduct-as-cast' format. The resulting polymers have been evaluated for use as separation media in capillary electrochromatography (CEC). The results have shown that substituting a small percentage of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service