Skip to Content
Merck
All Photos(2)

Key Documents

W255611

Sigma-Aldrich

γ-Hexalactone

natural, 97%, FG

Synonym(s):

γ-Caprolactone, γ-Ethyl-γ-butyrolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
FEMA Number:
2556
Beilstein:
6114822
EC Number:
Council of Europe no.:
2254
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.021
NACRES:
NA.21

grade

FG
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

97%

refractive index

n20/D 1.439 (lit.)

bp

219 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; herbaceous; sweet

SMILES string

CCC1CCC(=O)O1

InChI

1S/C6H10O2/c1-2-5-3-4-6(7)8-5/h5H,2-4H2,1H3

InChI key

JBFHTYHTHYHCDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Hexalactone is one the key volatile aroma compounds of peaches. It is also found in pawpaw fruit.
Natural Occurrence: Apricot, peach, mango, butter, cheees, milk, pawpaw, quince, papaya, raspberry, tea

Application

This chemical will deepen the fatty notes of nut flavors, especially pecan, macadamia, brazil nut, hazel nut and coconut It will add richness at low levels in fat flavors and replacers like tallow, milk fat and lard. Like other lactones, it will be useful in dairy flavors like cream, milk and cheese, and sweet brown flavors, especially caramel, toffee, vanilla and chocolate.

Biochem/physiol Actions

Taste at 5-10ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of key odor volatile compounds in the essential oil of nine peach accessions.
Eduardo I, et al.
Journal of the Science of Food and Agriculture, 90(7), 1146-1154 (2010)
Evaluation of headspace volatiles and sensory characteristics of ripe pawpaws (Asimina triloba) from selected cultivars
McGrath MJ & Karahadian C.
Food Chemistry, 51(3), 255-262 (1994)
Judith Michalski et al.
Perfumer & Flavorist, 39(2), 48-49 (2014)
H Thoma et al.
Journal of chromatography, 309(1), 17-32 (1984-07-13)
A two-step ultrafiltration method combined with anion-exchange chromatography is described for the separation of lipophilic and strongly lipophobic acids occurring in human blood, serum and plasma. After treatment with diazomethane, the acid fractions are separated further by gas chromatography. The
Olga Khersonsky et al.
Biochemistry, 44(16), 6371-6382 (2005-04-20)
PON1 is the best-studied member of a family of enzymes called serum paraoxonases, or PONs, identified in mammals (including humans) and other vertebrates as well as in invertebrates. PONs exhibit a range of important activities, including drug metabolism and detoxification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service