Skip to Content
Merck
All Photos(1)

Key Documents

W246000

Sigma-Aldrich

Ethyl tiglate

≥98%, FG

Synonym(s):

Ethyl trans-2-methyl-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2C2H5
CAS Number:
Molecular Weight:
128.17
FEMA Number:
2460
EC Number:
Council of Europe no.:
2185
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.495
NACRES:
NA.21
Organoleptic:
berry; caramel; fruity; tropical; floral; sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.435 (lit.)

bp

154-156 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; caramel; fruity; tropical; floral; sweet

SMILES string

CCOC(=O)\C(C)=C\C

InChI

1S/C7H12O2/c1-4-6(3)7(8)9-5-2/h4H,5H2,1-3H3/b6-4+

InChI key

OAPHLAAOJMTMLY-GQCTYLIASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl tiglate, an α,β-unsaturated ester, is naturally found in apples. It can be prepared by the esterification of tiglic acid. Ethyl tiglate is a male-specific pheromone found in some Drosophila species that can attract both male and females. Saprochaete suaveolens grown in a culture medium has been reported to produce ethyl tiglate by metabolizing isoleucine.

Application


  • Metabolism of ethyl tiglate in apple fruits leads to the formation of small amounts of (R)-ethyl 2-methylbutanoate.: Focuses on the metabolic transformation of ethyl tiglate in apples, which contributes to the fruit′s flavor profile (Hauck et al., 2000).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation of tiglic and angelic acids and esters1.
Buckles RE and Mock GV.
The Journal of Organic Chemistry, 15(3), 680-684 (1950)
Eric Grondin et al.
Yeast (Chichester, England), 32(1), 57-66 (2014-11-20)
A yeast identified as Saprochaete suaveolens was investigated for its capacity to produce a large panel of flavouring molecules. With a production of 32 compounds including 28 esters, S. suaveolens seems to be a good producer of fruity flavours and
R J Bartelt et al.
Journal of chemical ecology, 14(4), 1319-1327 (1988-04-01)
Mature males ofDrosophila borealis andD. littoralis (Diptera: Drosophilidae) produce pheromones that attract both males and females in a wind-tunnel bioassay. Ethyl tiglate is a major pheromone component in both species. Isopropyl tiglate is a minor component, as active as ethyl
Aggregation pheromones in five taxa of the Drosophila virilis species group.
Bartelt RJ, et al.
Physiological Entomology, 11(4), 367-376 (1986)
T Hauck et al.
Enantiomer, 5(5), 505-512 (2001-01-06)
(S)-Ethyl 2-methylbutanoate is an important aroma compound in apples and serves as an indicator for genuineness of apple products due to its high optical purity of greater than 98% enantiomeric excess [T. Koenig and P. Schreier, Zeitsch. Lebensm.-Unters. Forsch. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service