Skip to Content
Merck
All Photos(1)

Key Documents

W237108

Sigma-Aldrich

Benzyl ether

≥98%, FCC, FG

Synonym(s):

Dibenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
FEMA Number:
2371
Beilstein:
1911156
EC Number:
Council of Europe no.:
2150
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
3.004
NACRES:
NA.21
Organoleptic:
fruity; floral; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.561 (lit.)

bp

298 °C (lit.)

mp

1.5-3.5 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

dibenzyl ether

Organoleptic

fruity; floral; sweet

SMILES string

C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H14O/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

MHDVGSVTJDSBDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dibenzyl ether is a flavor and fragrance volatile used in the food and fragrance industry.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives, 1, 806-807 (1997)
Hui-Ling Dai et al.
Chemical & pharmaceutical bulletin, 57(1), 84-86 (2009-01-06)
Ten single benzyl phenyl ethers were synthesized and evaluated as human immunodeficiency virus-1 (HIV-1) inhibitors in vitro for the first time. Among these compounds, especially 4-nitrobenzyl phenyl ether (3h) exhibited the highest anti-HIV-1 activity with EC50 (concentration of drug that
Klaus Becker et al.
PloS one, 7(3), e33916-e33916 (2012-04-06)
Generally, chemical tissue clearing is performed by a solution consisting of two parts benzyl benzoate and one part benzyl alcohol. However, prolonged exposure to this mixture markedly reduces the fluorescence of GFP expressing specimens, so that one has to compromise
Ya-Ping Tu
The Journal of organic chemistry, 71(15), 5482-5488 (2006-07-15)
It is often found in mass spectrometry that when a molecule is protonated at the thermodynamically most favorable site, no fragmentation occurs, but a major reaction is observed when the proton migrates to a different position. For benzophenones, acetophenones, and
Hosamani Basavaprabhu et al.
Organic & biomolecular chemistry, 10(13), 2528-2533 (2012-03-01)
An application of the classical Ritter reaction for the synthesis of unsymmetrical di and trisubstituted ureas catalyzed by FeCl(3) is described. The protocol is of significant interest in view of the easy availability of precursors, mild reaction conditions employed and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service