Skip to Content
Merck
All Photos(1)

Key Documents

W236608

Sigma-Aldrich

trans-2-Decenal

≥95%, FCC, FG

Synonym(s):

(E)-2-decenal, (E)-3-heptyl acrolein

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH=CHCHO
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2366
Beilstein:
1704445
EC Number:
Council of Europe no.:
2009
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.191
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.453 (lit.)

bp

78-80 °C/3 mmHg (lit.)

density

0.841 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange; floral

SMILES string

[H]C(=O)\C([H])=C(/[H])CCCCCCC

InChI

1S/C10H18O/c1-2-3-4-5-6-7-8-9-10-11/h8-10H,2-7H2,1H3/b9-8+

InChI key

MMFCJPPRCYDLLZ-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-Decenal (C10 aldehyde) is an unsaturated trans-2 aldehyde found in the essential oil of coriander, bitter orange peel, ginger, butter, mushroom, and animal food. It can be used as a flavoring agent in food industries.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor Bites: trans-2-Decenal
WRIGHT J
Perfumer & Flavorist, 35, 34-35 (2010)
Optimization of photosynthetic photon flux density and root-zone temperature for enhancing secondary metabolite accumulation and production of coriander in plant factory
Nguyen DTP, et al.
advanced synthesis and catalysis, 9, 224-224 (2019)
Optimization of photosynthetic photon flux density and root-zone temperature for enhancing secondary metabolite accumulation and production of coriander in plant factory
Nguyen DTP, et al.
Agronomy, 9, 224-224 (2019)
Ahu Demirtas et al.
Anaerobe, 55, 83-92 (2018-11-06)
The effects of plant metabolites on rumen metabolism vary greatly depending on their antimicrobial spectrum and applied doses. In this study, the minimum inhibitory concentrations (MICs) of commercial aldehydes, trans-2-hexenal (T2H), cis-3-hexenal (C3H), trans-2-nonenal (T2N), and trans-2-decenal (T2D) from green

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service