Key Documents
TEM-G2000HACU
Sigma-Aldrich
2000 Mesh Circular Mesh Copper Grid
vial of 10
Synonym(s):
G2000HA CIRCULAR MESH COPPER GRIDS 10/VIAL, HEXAGONAL MESH 2000#
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
packaging
vial of 10
General description
High Mesh Value Standard Circular Mesh Grids: Copper Grid, 2000 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 12.5 μm, hole ~6.5μm, bar ~6μm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)