Key Documents
TEM-FF300CU
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 300 mesh, box of 25
Synonym(s):
FORMVAR FILM 300# COPPER25/PK
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
packaging
box of 25
size
300 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 300 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)