Skip to Content
Merck
All Photos(3)

Key Documents

T80489

Sigma-Aldrich

Trimethylsulfonium iodide

98%

Synonym(s):

Trimethyl-λ[3]-sulfane hydroiodide, Trimethylsulphonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3S(I)
CAS Number:
Molecular Weight:
204.07
Beilstein:
3555192
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

215-220 °C (lit.)

SMILES string

[I-].C[S+](C)C

InChI

1S/C3H9S.HI/c1-4(2)3;/h1-3H3;1H/q+1;/p-1

InChI key

VFJYIHQDILEQNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylsulfonium iodide is commonly used as a methylating agent in organic synthesis.

Application

Treatment with strong base yields the dimethylsulfonium methylide which reacts in-situ with the carbonyl group of ketones to form epoxides or allylic alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Díez et al.
Journal of chromatography. A, 1125(2), 244-253 (2006-06-20)
In this study, an orthogonal array design was applied to know the way different parameters affected the derivatization of some herbicides that are commonly applied in the soils. Herbicides formulated as esters have been reported to rapidly hydrolyse, in contact
Tetrahedron Letters, 35, 2009-2009 (1994)
Jörg Schönherr
Pest management science, 58(4), 343-351 (2002-04-27)
Penetration of glyphosate salts across isolated poplar (Populus canescens (Aiton) Sm) cuticular membranes (CM) was studied using Na+, K+, NH4+, trimethylsulfonium+ (TMS) and isopropylamine+ (IPA) as cations. After droplet drying, humidity over the salt residues on the outer surfaces of
Yasuyuki Ishida et al.
Journal of chromatography. A, 1216(15), 3296-3299 (2009-02-19)
Reaction efficiencies of two organic alkalis, tetramethylammonium hydroxide (TMAH) and trimethylsulfonium hydroxide (TMSH), with lipids during thermally assisted hydrolysis and methylation (THM) were examined focusing on (1) the types of lipids and (2) degree of unsaturation of fatty acid moieties.
K Müller et al.
Journal of clinical microbiology, 36(9), 2477-2480 (1998-08-15)
The MIDI automated Microbial Identification System (MIS) uses gas chromatography (GC) analysis of whole-cell fatty acid methyl esters (FAMEs) between 9 and 20 carbons in length to characterize a wide range of bacterial genera and species, including mycobacteria. Mycolic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service