Skip to Content
Merck
All Photos(2)

Key Documents

T35203

Sigma-Aldrich

trans-2,3-Dimethylacrylic acid

98%

Synonym(s):

Tiglic acid, trans-2,3-Dimethylacrylic acid, trans-2-Methyl-2-butenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)COOH
CAS Number:
Molecular Weight:
100.12
Beilstein:
1236500
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

95-96 °C/12 mmHg (lit.)

mp

61-64 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C=C(/C)C(O)=O

InChI

1S/C5H8O2/c1-3-4(2)5(6)7/h3H,1-2H3,(H,6,7)/b4-3+

InChI key

UIERETOOQGIECD-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

203.0 °F

Flash Point(C)

95 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K D Santos et al.
International journal of immunopharmacology, 18(12), 761-769 (1996-12-01)
The present study was undertaken to assess the influence of 25 organic acids, which appear in high concentrations in tissues of patients with various organic acidaemias, on the proliferation of human peripheral lymphocytes stimulated with concanavalin A (Con A) (a
Tigliane-type diterpene esters from Synadenium grantii.
R Bagavathi et al.
Planta medica, 54(6), 506-510 (1988-12-01)
Araceli Larios et al.
Applied microbiology and biotechnology, 65(4), 373-376 (2004-07-13)
Candida antarctica lipase fraction B (CAL-B) showed substrate specificity in the synthesis of esters in hexane involving reactions of short-chain acids having linear (acetic and butyric acids) and branched chain (isovaleric acid) structures, an unsaturated (tiglic acid) fatty acid, and
[Effects of a skin stimulating salve].
W RIESE
Wiener medizinische Wochenschrift (1946), 100(45-46), 758-759 (1950-11-25)
Susan Matthew et al.
Phytochemistry, 70(17-18), 2058-2063 (2009-10-10)
The Floridian marine cyanobacterium Lyngbya confervoides afforded cyclodepsipeptides, termed tiglicamides A-C (1-3), along with their previously reported analogues largamides A-C (4-6), all of which possess an unusual tiglic acid moiety. Their structures were deduced by one- and two-dimensional NMR combined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service