Skip to Content
Merck
All Photos(4)

Key Documents

T33200

Sigma-Aldrich

Thiosalicylic acid

97%

Synonym(s):

2-Sulfanylbenzoic acid, 2-Mercaptobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4CO2H
CAS Number:
Molecular Weight:
154.19
Beilstein:
508507
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

162-165 °C (lit.)

SMILES string

OC(=O)c1ccccc1S

InChI

1S/C7H6O2S/c8-7(9)5-3-1-2-4-6(5)10/h1-4,10H,(H,8,9)

InChI key

NBOMNTLFRHMDEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thiosalicylic acid can be used as:
  • A nucleophilic trapping agent for the desulfenylation of 3-indolyl sulfides to obtain 3-unsubstituted indoles.
  • A starting material to prepare 2′-mercaptoacetophenone, which is used in the synthesis of thioflavanone by reacting with lithium diisopropylamide and benzaldehyde.
  • A stabilizing agent in the synthesis of metal nanoparticles.

It can also be used to prepare 2-thioxanthone-thioacetic acid bimolecular system, which is used as a photoinitiator for free radical polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, 309-309 (1994)
R Grossman et al.
NeuroImage, 20(4), 1971-1981 (2003-12-20)
Increases in peripheral type benzodiazepine receptors (PTBR) have been utilized for the detection of neuroinflammation and neurotoxicity in the brain. We have investigated the relationship between PTBR and NMDA receptor binding density in mice with closed head injury (CHI) using
Elisabet Bermudo et al.
Journal of chromatography. A, 1129(1), 129-134 (2006-07-18)
Two in-line preconcentration capillary zone electrophoresis (CZE) methods (field amplified sample injection (FASI) and stacking with sample matrix removal (LVSS)) have been evaluated for the analysis of acrylamide (AA) in foodstuffs. To allow the determination of AA by CZE, it
S Chong et al.
Biochemical pharmacology, 42(7), 1433-1439 (1991-09-12)
Co-administration of N-acetylcysteine (NAC) with nitroglycerin (NTG) has been shown to partially reverse nitrate tolerance and to potentiate the hypotensive effect of NTG in humans. However, a high clinical dose of NAC was required for this pharmacologic interaction resulting in
Synthesis and characterization of thiosalicylic acid stabilized gold nanoparticles
Pattabi RM and Pattabi M
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 74(1), 195-199 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service