T27804
4-(2-Thienyl)butyric acid
98%
Synonym(s):
2-Thiophenebutyric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H10O2S
CAS Number:
Molecular Weight:
170.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.532 (lit.)
density
1.169 g/mL at 25 °C (lit.)
SMILES string
OC(=O)CCCc1cccs1
InChI
1S/C8H10O2S/c9-8(10)5-1-3-7-4-2-6-11-7/h2,4,6H,1,3,5H2,(H,9,10)
InChI key
VYTXLSQVYGNWLV-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
L H Lash et al.
Toxicology, 103(3), 167-175 (1995-12-10)
Relative cytotoxicity and mechanism of action of thiophenes were investigated in suspensions of freshly isolated renal proximal tubular (PT) and distal tubular (DT) cells using 4-(2-thienyl)butyric acid (TByA) as a model compound. TByA produced significantly greater time- and concentration-dependent cytotoxicity
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service