Skip to Content
Merck
All Photos(3)

Key Documents

T27502

Sigma-Aldrich

L-4-Thiazolidinecarboxylic acid

98%

Synonym(s):

L-Thiaproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2S
CAS Number:
Molecular Weight:
133.17
Beilstein:
81065
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

optical activity

[α]20/D −141°, c = 1.3 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

190-200 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1CSCN1

InChI

1S/C4H7NO2S/c6-4(7)3-1-8-2-5-3/h3,5H,1-2H2,(H,6,7)/t3-/m0/s1

InChI key

DZLNHFMRPBPULJ-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in peptide coupling reactions.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioorganic & Medicinal Chemistry Letters, 4, 887-887 (1994)
Florine Cavelier et al.
Journal of peptide science : an official publication of the European Peptide Society, 12(10), 621-625 (2006-06-21)
As proline plays an important role in biologically active peptides, many analogues of this residue have been developed to modulate the proportion of cis and trans conformers. A correlation between the pyrrolidine ring shape and structural properties of proline has
Masashi Suo et al.
Cancer letters, 237(2), 256-262 (2005-07-30)
An excessive duodenal reflux induced by surgery has been widely accepted to cause gastric carcinogenesis in the remnant stomach. As one of causative factors for malignancy, N-nitroso compounds produced by enteric bacteria have been postulated. However, there is no concrete
Pedro P Corbi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 929-931 (2008-03-21)
A new water-soluble lithium salt of thiazolidine-4-carboxylic acid was synthesized and characterized by chemical and spectroscopic techniques. Elemental and mass spectrometric (ESI-MS) analyses of the solid compound fit to the composition LiC(4)H(6)NSO(2). (1)H, (13)C nuclear magnetic resonance (NMR), [(1)H-(15)N] NMR
Anahí Magdaleno et al.
PloS one, 4(2), e4534-e4534 (2009-02-21)
It is well established that L-proline has several roles in the biology of trypanosomatids. In Trypanosoma cruzi, the etiological agent of Chagas' disease, this amino acid is involved in energy metabolism, differentiation processes and resistance to osmotic stress. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service