Key Documents
SYNS580008
Synthware 3-way high vacuum valve
size 0-8 mm
Synonym(s):
Synthware S580008
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S580008
General description
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another. The non-rotating PTFE plug has three O-rings, the tip O-ring making the primary seal on the precision formed tapered seat.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)