SYNF534250
Synthware™ single neck reaction flask with glass stopcock
capacity 250 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F534250, Schlenk flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
round bottom
single-neck (with glass stopcock)
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F534250
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With 2 mm glass stopcock. With spring retention device for smooth turning purpose.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service