SYNF484008
Synthware™ solvent transfer/storage flask
1000 mL, joint: ST/NS 24/40, valve size 0-8 mm
Synonym(s):
Synthware F484008, Straus flask, receiver flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
round bottom
two-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F484008
capacity
1,000 mL
1000 mL
1000 mL
valve size
0-8 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Synthware™ solvent transfer/storage flask features a center neck joint sealed off from direct access to flask. A syringe or cannula is used to recover the solvent through the valve.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service