Skip to Content
Merck
All Photos(1)

Key Documents

SYNF434450

Synthware three neck round bottom flask with angled side necks

50 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40

Synonym(s):

Synthware F434450, 3-neck round flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.51

Pricing and availability is not currently available.

material

borosilicate glass flask
round bottom
three-neck (angled side necks)

Joint

center joint: ST/NS24/40
side joint: ST/NS24/40

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F434450

capacity

50 mL
50 mL

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF59743738GF67966385GF21271025
assay

≥99.7%

assay

99.7%

assay

99.7%

assay

99.70%

manufacturer/tradename

Goodfellow 957-976-92

manufacturer/tradename

Goodfellow 597-437-38

manufacturer/tradename

Goodfellow 679-663-85

manufacturer/tradename

Goodfellow 212-710-25

resistivity

4.46 μΩ-cm, 20°C

resistivity

4.46 μΩ-cm, 20°C

resistivity

4.46 μΩ-cm, 20°C

resistivity

4.46 μΩ-cm, 20°C

density

1.85 g/mL at 25 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

General description

Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.

Legal Information

Synthware is a trademark of Kemtech America Inc.

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service