Key Documents
SYND441405
Synthware Dean Stark distillation receiver with vacuum jacketed uptake tube
5 mL, joint: ST/NS 14/20
Synonym(s):
Synthware D441405, Dean-Stark apparatus, Distilling receiver, Dean-Stark trap
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D441405
capacity
5 mL
Related Categories
General description
Similar to D451414 above but with the uptake tube vacuum jacketed to improve distillation performance. The jacket eliminates the possibility of condensation or recrystallization of distillate in the sidearm. With 2 mm PTFE stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)