Key Documents
SYND171014
Synthware Vigreux vacuum jacketed micro distillation head
joint: ST/NS 14/20, thermometer joint: ST/NS 10/18, immersion depth 25 mm
Synonym(s):
Synthware D171014, Short path head
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Joint
thermometer joint: ST/NS10/18
joint: ST/NS14/20
manufacturer/tradename
D171014
immersion depth
25 mm
Related Categories
General description
Synthware distillation head features a Vigreux column jacketed to improve distillation performance. The standard taper joint (10/18) accommodates a thermometer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)