Key Documents
SYND121014
Synthware vacuum jacketed short path distillation head
joint: ST/NS 14/20, thermometer joint: ST/NS 10/18, immersion depth 25 mm
Synonym(s):
Synthware D121014, Distilling head
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
thermometer joint: ST/NS10/18
joint: ST/NS14/20
manufacturer/tradename
D121014
immersion depth
25 mm
Related Categories
General description
Synthware vacuum-jacketed design for enhanced efficiency. When a thermometer is inserted at the top of this apparatus, the bulb and stem act as column packing. The close-coupled condenser and vacuum connection provide an extremely short condensate travel path. This apparatus is compatible with a 25 mm immersion thermometer and a D27 distillation receiver.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)