Key Documents
SYNC564175
Synthware coil-style reflux condenser with removable hose connections
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C564175, coil-type condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C564175
hose size
10 mm
Related Categories
General description
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)