Key Documents
SYNC291910
Synthware graduated concentrator tube
10 mL, joint: ST/NS 19/22
Synonym(s):
Synthware C291910, glass concentrator tube
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available. Contact Technical Service
material
colorless tube
glass tube
Joint
joint: ST/NS19/22
Agency
suitable for EPA 1613
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C291910
capacity
10 mL
General description
Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups. Tube can be stoppered for storage. Overall height is 100 mm for 10 mL, and 170 mm for the 25 mL.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.