Key Documents
SYNC221420
Synthware air reflux condenser
joint: ST/NS 14/20, L 96 mm
Synonym(s):
Synthware C221420, Glass air condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pack of 1 ea
manufacturer/tradename
C221420
L
96 mm
Related Categories
General description
Synthware air reflux is a condenser enhancing separation through a reflux mechanism. With upper standard taper outer joint (14/20) to fit various laboratory glassware and a lower drip tip joint. Overall length is 143 mm and length between the joints is 96 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)