Key Documents
SYNC114250
Synthware cold finger condenser
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C114250, Cold finger condenser with drip tip
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C114250
hose size
10 mm
Related Categories
General description
Extended offset lower tip for drop control.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)