SYNA692440
Synthware™ 90 degree vacuum adapter with glass stopcock
female joint: ST/NS 24/40
Synonym(s):
Synthware A692440, Flow control adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
female joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A692440
Related Categories
General description
For regulating the input of gases or liquids into reaction systems. With 8 mm or 10 mm hose connection. With 8 mm or 10 mm hose connection.
Legal Information
Synthware is a trademark of Kemtech America Inc.