Key Documents
SYNA412440
Synthware distillation drip adapter
joint: ST/NS 24/40, Bend: 105 degree
Synonym(s):
Synthware A412440, bent distillation adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pack of 1 ea
manufacturer/tradename
A412440
Related Categories
General description
Synthware distillation drop adapter with standard joint (24/40). To effectively clean the rotary evaporator.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)