SM4050105
SmartPor, alumina membrane
thickness 50 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Al
UNSPSC Code:
12352103
NACRES:
NA.23
Recommended Products
description
Pore geometry: trigonal arrangement
packaging
pkg of 5 pieces
size
10 mm × 10 mm
thickness
50 μm
pore size
125 nm (interpore distance)
40 nm pore diameter
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service