Key Documents
SLW5809/3
Pyrex® Filter holder for 47 mm membranes
base only
Synonym(s):
Pyrex, SciLabware 5809/3
About This Item
UNSPSC Code:
41123300
NACRES:
NB.43
packaging
pkg of 1 ea
manufacturer/tradename
5809/3
SciLabware
General description
Comprising of a 300 ml capacity funnel, support base and metal clamp, this popular size filter holder is designed to be used with 47 mm membrane filters. The three components of this product must be ordered separately.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)