SLW3720/02M
Pyrex® Büchner filter funnels with sintered disc
1000 mL, disc diam. 120 mm, porosity grade 1
Synonym(s):
Pyrex, SciLabware 3720/02M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
manufacturer/tradename
3720/02M
SciLabware
capacity
1000 mL
1000 mL
disc diam.
120 mm
pore size
grade 1 porosity
Looking for similar products? Visit Product Comparison Guide
General description
The 120mm sintered discs fitted to these funnels are manufactured from the same 3.3 expansion borosilicate glass as the funnel bodies, making these funnels suitable to be used with a wide range aggressive chemicals.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service