SLW3215/06MP
Pyrex® Mixing cylinder, class B, detachable foot,
25 mL
Synonym(s):
Pyrex, SciLabware 3215/06MP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polyethylene base
polyethylene stopper
Joint
joint stopper14/23
feature
graduations 0.5 mL
detachable foot
manufacturer/tradename
3215/06MP
SciLabware
H
175 mm
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with ISO 4788
- Comprises borosilicate glass cylinder, polyethylene foot, and polyethylene stopper
- Replacement parts available separately
- Graduations in white enamel
Features and Benefits
• Complies with ISO 4788• Manufactured from borosilicate glass 3.3• Calibrated to Class B tolerances• With polyethylene foot and polyethylene stopper• Graduations in white enamel• Replacement parts available separately
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service