SLW3151/20WC
Pyrex® Volumetric flask, heavy duty, class A, works certified, USP/ISO/DIN tolerances
capacity 500 mL
Synonym(s):
Pyrex, SciLabware 3151/20WC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass flask
Joint
joint ST/NS neck19/26
feature
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
packaging
pkg of 2 ea
manufacturer/tradename
3151/20WC
SciLabware
Overall H
260 mm
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Class A, works certified
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerances required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
- Each flask engraved with individual serial number and supplied with certificate of calibration traceable to National standards
- Conformity ‘H′ certified
- Manufactured from borosilicate glass for corrosion-free performance
- White enamel inscriptions
- Polyethylene stopper
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service