SIK7509-10
11-(2-Bromoisobutyrate)-undecyl-1-phosphonic acid
≥95%
About This Item
Recommended Products
Assay
≥95%
form
solid
shelf life
3 years under the recommended conditions
color
white
SMILES string
BrC(C)(C(OCCCCCCCCCCCP(O)(O)=O)=O)C
InChI
1S/C15H30BrO5P/c1-15(2,16)14(17)21-12-10-8-6-4-3-5-7-9-11-13-22(18,19)20/h3-13H2,1-2H3,(H2,18,19,20)
InChI key
SBFNQWAMVLRANY-UHFFFAOYSA-N
Related Categories
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service