All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H15O4P
CAS Number:
Molecular Weight:
182.15
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.23
Recommended Products
Assay
97%
form
powder
shelf life
3 years under the recommended conditions
color
white
SMILES string
[P](=O)(O)(O)CCCCCCO
InChI
1S/C6H15O4P/c7-5-3-1-2-4-6-11(8,9)10/h7H,1-6H2,(H2,8,9,10)
InChI key
NVQCNUFHCNCGKU-UHFFFAOYSA-N
Related Categories
Application
These products are used as linkers to improve adhesion between coatings and metal surfaces. The phosphonic acid groups can graft onto metal surface while the hydroxyl groups react with common epoxy/amine-based paints.
Legal Information
Product of SiKEMIA
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service