QBD10910
m-dPEG®37-NHS ester
>95% (HPLC)
Synonym(s):
m-PEG37-NHS ester, mPEG-NHS ester, mPEG1500-NHS ester, mPEG37-NHS ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C80H155NO41
Molecular Weight:
1787.07
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22
Recommended Products
Assay
>95% (HPLC)
form
solid or viscous liquid
reaction suitability
reaction type: Pegylations
polymer architecture
shape: linear
functionality: monofunctional
shipped in
ambient
storage temp.
−20°C
Features and Benefits
m-dPEG37-NHS ester is a methyl-terminated, single molecular weight, discrete chain length polyethylene glycol (dPEG) spacer. The carboxylic acid moiety on the end of the molecule opposite the methyl cap is activated with N-hydroxysuccinimide (NHS) for facile reaction with free amines. The dPEG spacer is 112 atoms (133.9 Å) long, a monodisperse equivalent of a polymeric PEG1500. m-dPEG37-NHS ester can be used to modify silica shelled quantum dots to make them water-soluble. Also, it has been employed to improve the pharmacokinetic (PK) and biodistribution (BD) properties of an antibody. Carbon nanotubes and inorganic nanoparticles with amine-functionalized surfaces could also be modified to passivate the surface and make it hydrophilic. Overall, proteins and other biomolecules conjugated to this product will have higher water solubility, reduced or eliminated antigenicity, and improved PK and BD properties.
Legal Information
Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service