Skip to Content
Merck
All Photos(1)

Key Documents

QBD10908

Sigma-Aldrich

m-dPEG®36-amine

>95% (HPLC)

Synonym(s):

m-PEG36-amine, Methyl-PEG36-amine, mPEG-amine, mPEG1500-amine, mPEG36-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H149NO36
Molecular Weight:
1616.95
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG36-amine is a methyl-terminated PEG amine compound. The PEG is a water-soluble, single molecular weight PEG product with a discrete chain length (dPEG). This dPEG linker is 109 atoms (130.0 Å) long. Carboxylic acids and their active esters react with the amino terminus to form amide bonds. Also, the amino terminus reacts with aldehydes and ketones, forming Schiff bonds, which are reducible to secondary amines. m-dPEG36-amine is a monodisperse equivalent of the polymeric PEG1500. Uses for m-dPEG36-amine include modification of acid-functionalized surfaces and free carboxylic acid groups on biomolecules.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service