Skip to Content
Merck
All Photos(1)

Key Documents

QBD10774

Sigma-Aldrich

NHS-dPEG®24-biotin

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H120N4O30S
Molecular Weight:
1469.72
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reactivity: amine reactive

reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCC1SCC(N2)C1NC2=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON3C(CCC3=O)=O)=O

General description

NHS-dPEG®24-biotin is a cross-linking reagent used in biotinylation reactions. The N-hydroxysuccinimidyl (NHS) moiety of NHS-dPEG®24-biotin forms a stable amide bond with free primary amines, such as the amines on the side chain of lysine. The biotin moiety is used in a variety of applications, including labeling, supramolecular building, affinity chromatography, pull-down assays, and the production of biotinylated antibodies.

Features and Benefits

NHS-dPEG®24-biotin is used extensively to label molecules that contain a free amine with biotin. The single molecular weight NHS-dPEG®24-biotin compound has a long spacer of precise length (97.7 Å) between the biotin moiety and the labeled molecule. This dPEG® spacer imparts a high degree of water solubility to the biotin label. For biotinylated peptides and proteins, the dPEG® spacer dramatically reduces aggregation and precipitation of the modified molecule and reduces non-specific binding of the molecule, both of which are common problems with molecules that have been biotinylated with the widely used NHS-LC-biotin.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic separation and enzymatic catalysis conjugated colorimetric immunosensor for Hepatitis B surface antigen detection
Hu Yongqin, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 168 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service